Gene Transcripts

Gene Symbol
Rell2
Entrez Gene ID
361313
Gene Name
RELT-like 2
Gene Aliases
RGD1305276
Chromosome Location
Chr. 18: 31094965 - 31098536 on Build Rnor_6.0
UniGene ID
Rn.145169
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014149.1 NP_001014171.1 6 1218
GenBank mRNA BC089946.1 - 6 1218

View Full Product Details