Gene Transcripts
- Gene Symbol
- RGS9
- Entrez Gene ID
- 8787
- Gene Name
- regulator of G-protein signaling 9
- Gene Aliases
- PERRS, RGS9L
- Chromosome Location
- Chr. 17: 65137338 - 65227703 on Build GRCh38
- UniGene ID
- Hs.664380
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001081955.2 |
NP_001075424.1
|
10, 11 |
885 |
|
NM_001165933.1 |
NP_001159405.1
|
10, 11 |
885 |
|
NM_003835.3 |
NP_003826.2
|
10, 11 |
894 |
|
XM_011525426.2 |
XP_011523728.1
|
3, 4 |
1065 |
|
GenBank mRNA |
AF071476.1 |
- |
10, 11 |
758 |
|
AF073710.1 |
- |
1, 2 |
619 |
|
AF493933.1 |
- |
10, 11 |
691 |
|
AK289876.1 |
- |
10, 11 |
758 |
|
AK290535.1 |
- |
10, 11 |
754 |
|
AK301719.1 |
- |
10, 11 |
764 |
|
AK309061.1 |
- |
10, 11 |
775 |
|
AY585191.1 |
- |
10, 11 |
682 |
|
BC022504.1 |
- |
10, 11 |
752 |
4801328 |
View Full Product Details