Gene Transcripts

Gene Symbol
Etv2
Entrez Gene ID
361544
Gene Name
ets variant 2
Gene Aliases
Etsrp71
Chromosome Location
Chr. 1: 89091239 - 89094570 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006228814.2 XP_006228876.1 7 1769
XM_006228815.2 XP_006228877.1 7 1787
XM_008759255.1 XP_008757477.1 6 1990

View Full Product Details