Gene Transcripts

Gene Symbol
Lemd2
Entrez Gene ID
361807
Gene Name
LEM domain containing 2
Gene Aliases
Chromosome Location
Chr. 20: 6545083 - 6556350 on Build Rnor_6.0
UniGene ID
Rn.14675
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039032.1 NP_001034121.1 1 249
GenBank mRNA BC098775.1 - 1 249

View Full Product Details