Gene Transcripts

Gene Symbol
BANF1
Entrez Gene ID
8815
Gene Name
barrier to autointegration factor 1
Gene Aliases
BAF, BCRP1, D14S1460, NGPS
Chromosome Location
Chr. 11: 66002079 - 66004149 on Build GRCh38
UniGene ID
Hs.433759
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143985.1 NP_001137457.1 3 816
NM_003860.3 NP_003851.1 3 873
XM_017018514.1 XP_016874003.1 3 866
XM_017018515.1 XP_016874004.1 3 406
GenBank mRNA AF044773.1 - 3 466
AF068235.1 - 3 872
BC005942.1 - 3 452 4074168
BC107702.1 - 3 549 6495215
BG748852.1 - 3 657
BU940708.1 - 3 633

View Full Product Details