Gene Transcripts

Gene Symbol
Cdc40
Entrez Gene ID
361859
Gene Name
cell division cycle 40
Gene Aliases
Chromosome Location
Chr. 20: 48503973 - 48574546 on Build Rnor_6.0
UniGene ID
Rn.14862
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108538.1 NP_001102008.1 10 1302
XM_008773000.1 XP_008771222.1 12 1272
GenBank mRNA BC166724.1 - 12 1233

View Full Product Details