Gene Transcripts

Gene Symbol
SAP30
Entrez Gene ID
8819
Gene Name
Sin3A associated protein 30
Gene Aliases
Chromosome Location
Chr. 4: 173370942 - 173377532 on Build GRCh38
UniGene ID
Hs.591715
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003864.3 NP_003855.1 3, 4 774
GenBank mRNA AF055993.1 - 3, 4 773
BC016757.1 - 3, 4 769 4074154

View Full Product Details