Gene Transcripts

Gene Symbol
Slc39a1
Entrez Gene ID
361986
Gene Name
solute carrier family 39 (zinc transporter), member 1
Gene Aliases
Chromosome Location
Chr. 2: 189609766 - 189615367 on Build Rnor_6.0
UniGene ID
Rn.43952
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134577.1 NP_001128049.1 4 816
XM_006232707.2 XP_006232769.1 4 864
GenBank mRNA BC168649.1 - 4 816

View Full Product Details