Gene Transcripts

Gene Symbol
Rapsn
Entrez Gene ID
362161
Gene Name
receptor-associated protein of the synapse
Gene Aliases
Chromosome Location
Chr. 3: 79859816 - 79869486 on Build Rnor_6.0
UniGene ID
Rn.17149
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108584.1 NP_001102054.1 2 377
XM_006234528.2 XP_006234590.1 2 768

View Full Product Details