Gene Transcripts

Gene Symbol
Mapkbp1
Entrez Gene ID
362197
Gene Name
mitogen activated protein kinase binding protein 1
Gene Aliases
Chromosome Location
Chr. 3: 111767126 - 111817101 on Build Rnor_6.0
UniGene ID
Rn.100547
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108589.1 NP_001102059.1 16 1970
XM_006234781.2 XP_006234843.1 15 2327

View Full Product Details