Gene Transcripts
- Gene Symbol
- FCGBP
- Entrez Gene ID
- 8857
- Gene Name
- Fc fragment of IgG binding protein
- Gene Aliases
- FC(GAMMA)BP
- Chromosome Location
- Chr. 19: 39863323 - 39934626 on Build GRCh38
- UniGene ID
- Hs.111732
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003890.2 |
NP_003881.2
|
|
3196 |
|
GenBank mRNA |
D84239.1 |
- |
|
3196 |
|
View Full Product Details