Gene Transcripts

Gene Symbol
Ndufaf4
Entrez Gene ID
362495
Gene Name
NADH dehydrogenase (ubiquinone) complex I, assembly factor 4
Gene Aliases
Hrpap20
Chromosome Location
Chr. 5: 39251370 - 39256542 on Build Rnor_6.0
UniGene ID
Rn.106694
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198783.1 NP_942078.1 3 377
GenBank mRNA AF283900.1 - 3 377
AF283901.1 - 2 640

View Full Product Details