Gene Transcripts

Gene Symbol
Trit1
Entrez Gene ID
362586
Gene Name
tRNA isopentenyltransferase 1
Gene Aliases
Chromosome Location
Chr. 5: 140712002 - 140756088 on Build Rnor_6.0
UniGene ID
Rn.74419
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108676.1 NP_001102146.1 4 497
XM_006238804.2 XP_006238866.1 4 1036

View Full Product Details