Gene Transcripts

Gene Symbol
Fbxo42
Entrez Gene ID
362646
Gene Name
F-box protein 42
Gene Aliases
Chromosome Location
Chr. 5: 159670662 - 159729070 on Build Rnor_6.0
UniGene ID
Rn.162839
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108691.1 NP_001102161.1 10 1685
XM_006239253.2 XP_006239315.1 10 1780

View Full Product Details