Gene Transcripts

Gene Symbol
Thap3
Entrez Gene ID
362667
Gene Name
THAP domain containing, apoptosis associated protein 3
Gene Aliases
RGD1308109
Chromosome Location
Chr. 5: 169155239 - 169160987 on Build Rnor_6.0
UniGene ID
Rn.14849
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108695.1 NP_001102165.1 1, 2 192
GenBank mRNA BC159427.1 - 1, 2 90
FQ229895.1 - 1, 2 78

View Full Product Details