Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001108702.2 | NP_001102172.2 | 5 | 873 | |
XM_008764486.1 | XP_008762708.1 | 6 | 1249 | ||
XM_008764487.1 | XP_008762709.1 | 6 | 1246 | ||
XM_008764488.1 | XP_008762710.1 | 5 | 1154 | ||
GenBank mRNA | BC166846.1 | - | 5 | 873 |