Gene Transcripts

Gene Symbol
Ilvbl
Entrez Gene ID
362843
Gene Name
ilvB (bacterial acetolactate synthase)-like
Gene Aliases
Chromosome Location
Chr. 7: 14044586 - 14054639 on Build Rnor_6.0
UniGene ID
Rn.22332
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108738.1 NP_001102208.1 14, 15 2096
XM_006241070.2 XP_006241132.1 14, 15 1753
XM_006241071.2 XP_006241133.1 14, 15 1748

View Full Product Details