Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_003750698.3 | XP_003750746.1 | 4 | 4535 | |
XM_006244936.2 | XP_006244998.1 | 4 | 1471 | ||
XM_006244937.2 | XP_006244999.1 | 5 | 1459 | ||
XM_006244938.2 | XP_006245000.1 | 4 | 2489 | ||
XM_008767152.1 | XP_008765374.1 | 4 | 2047 |