Gene Transcripts

Gene Symbol
Cops7b
Entrez Gene ID
363273
Gene Name
COP9 signalosome subunit 7B
Gene Aliases
Chromosome Location
Chr. 9: 93607171 - 93633352 on Build Rnor_6.0
UniGene ID
Rn.33834
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108807.1 NP_001102277.1 6 773
XM_006245414.2 XP_006245476.1 5 1224
XM_006245415.2 XP_006245477.1 5 861

View Full Product Details