Gene Transcripts

Gene Symbol
Chmp2b
Entrez Gene ID
363720
Gene Name
charged multivesicular body protein 2B
Gene Aliases
RGD1306781
Chromosome Location
Chr. 11: 2665264 - 2692254 on Build Rnor_6.0
UniGene ID
Rn.105547
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_343995.7 XP_343996.5 5 776
GenBank mRNA FQ212834.1 - 5 741

View Full Product Details