Gene Transcripts

Gene Symbol
Ccdc94
Entrez Gene ID
501285
Gene Name
coiled-coil domain containing 94
Gene Aliases
Chromosome Location
Chr. 9: 11098914 - 11108805 on Build Rnor_6.0
UniGene ID
Rn.198440
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109673.1 NP_001103143.1 5 498
XM_006244336.2 XP_006244398.1 5 562
GenBank mRNA BC129098.1 - 5 513
BC169032.1 - 5 498 9091720

View Full Product Details