Gene Transcripts
- Gene Symbol
- CDKL2
- Entrez Gene ID
- 8999
- Gene Name
- cyclin dependent kinase like 2
- Gene Aliases
- KKIAMRE, P56
- Chromosome Location
- Chr. 4: 75576494 - 75630537 on Build GRCh38
- UniGene ID
- Hs.591698
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003948.3 |
NP_003939.1
|
5, 6 |
1181 |
|
XM_005263335.2 |
XP_005263392.1
|
5, 6 |
900 |
|
XM_006714406.2 |
XP_006714469.1
|
5, 6 |
900 |
|
XM_017008809.1 |
XP_016864298.1
|
5, 6 |
900 |
|
XM_017008810.1 |
XP_016864299.1
|
5, 6 |
900 |
|
XM_017008811.1 |
XP_016864300.1
|
5, 6 |
900 |
|
GenBank mRNA |
AK294866.1 |
- |
5, 6 |
1167 |
|
AK312490.1 |
- |
5, 6 |
1172 |
|
BC093646.1 |
- |
5, 6 |
705 |
7939491 |
BC093981.1 |
- |
5, 6 |
705 |
7939826 |
U35146.1 |
- |
5, 6 |
1031 |
|
View Full Product Details