Gene Transcripts

Gene Symbol
Inpp5a
Entrez Gene ID
365382
Gene Name
inositol polyphosphate-5-phosphatase A
Gene Aliases
Chromosome Location
Chr. 1: 211732163 - 211920844 on Build Rnor_6.0
UniGene ID
Rn.8618
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108923.2 NP_001102393.1 8 839
XM_006230486.2 XP_006230548.1 8 1146

View Full Product Details