Gene Transcripts

Gene Symbol
Bhlhe22
Entrez Gene ID
365748
Gene Name
basic helix-loop-helix family, member e22
Gene Aliases
Bhlhb5
Chromosome Location
Chr. 2: 102685513 - 102688624 on Build Rnor_6.0
UniGene ID
Rn.161818
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108940.1 NP_001102410.1 1 1964

View Full Product Details