Gene Transcripts

Gene Symbol
Rab33b
Entrez Gene ID
365793
Gene Name
RAB33B, member RAS oncogene family
Gene Aliases
Chromosome Location
Chr. 2: 140541619 - 140552220 on Build Rnor_6.0
UniGene ID
Rn.17886
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108944.1 NP_001102414.1 1 517

View Full Product Details