Gene Transcripts

Gene Symbol
Tmco5b
Entrez Gene ID
366158
Gene Name
transmembrane and coiled-coil domains 5B
Gene Aliases
RGD1561237
Chromosome Location
Chr. 3: 104721643 - 104765412 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008762110.1 XP_008760332.1 5 422
XM_575205.5 XP_575205.2 5 366

View Full Product Details