Gene Transcripts

Gene Symbol
R3hdml
Entrez Gene ID
366245
Gene Name
R3H domain containing-like
Gene Aliases
Chromosome Location
Chr. 3: 159886248 - 159897993 on Build Rnor_6.0
UniGene ID
Rn.218529
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108962.1 NP_001102432.1 5 875
XM_006235600.2 XP_006235662.1 6 1018

View Full Product Details