Gene Transcripts

Gene Symbol
Cfl2
Entrez Gene ID
366624
Gene Name
cofilin 2, muscle
Gene Aliases
Chromosome Location
Chr. 6: 75759140 - 75763185 on Build Rnor_6.0
UniGene ID
Rn.101880
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108982.2 NP_001102452.2 2 153
XM_006240104.2 XP_006240166.1 2 240
GenBank mRNA FQ224178.1 - 2 83
FQ228857.1 - 2 160

View Full Product Details