Gene Transcripts

Gene Symbol
Tonsl
Entrez Gene ID
366953
Gene Name
tonsoku-like, DNA repair protein
Gene Aliases
Nfkbil2
Chromosome Location
Chr. 7: 117688397 - 117703130 on Build Rnor_6.0
UniGene ID
Rn.19092
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130572.1 NP_001124044.1 9 1185
XM_008765587.1 XP_008763809.1 9 1221
XM_008765588.1 XP_008763810.1 9 1221

View Full Product Details