Gene Transcripts

Gene Symbol
Kb15
Entrez Gene ID
366992
Gene Name
type II keratin Kb15
Gene Aliases
Chromosome Location
Chr. 7: 143208034 - 143217480 on Build Rnor_6.0
UniGene ID
Rn.49197
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008825.1 NP_001008825.1 6 1112
XM_006242347.2 XP_006242409.1 6 1182

View Full Product Details