Gene Transcripts

Gene Symbol
Zc4h2
Entrez Gene ID
367838
Gene Name
zinc finger, C4H2 domain containing
Gene Aliases
RGD1561708
Chromosome Location
Chr. X: 64887978 - 64908682 on Build Rnor_6.0
UniGene ID
Rn.12889
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001126374.1 NP_001119846.1 1, 2 165
GenBank mRNA BC162002.1 - 1, 2 165

View Full Product Details