Gene Transcripts
- Gene Symbol
- DNAJA3
- Entrez Gene ID
- 9093
- Gene Name
- DnaJ heat shock protein family (Hsp40) member A3
- Gene Aliases
- HCA57, TID1, hTID-1
- Chromosome Location
- Chr. 16: 4425805 - 4456775 on Build GRCh38
- UniGene ID
- Hs.459779
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001135110.2 |
NP_001128582.1
|
2, 3 |
415 |
|
NM_005147.5 |
NP_005138.3
|
2, 3 |
415 |
|
GenBank mRNA |
AB209923.1 |
- |
2, 3 |
334 |
|
AF061749.1 |
- |
2, 3 |
369 |
|
AF411044.1 |
- |
2, 3 |
382 |
|
AK001333.1 |
- |
2, 3 |
355 |
|
AK223105.1 |
- |
2, 3 |
355 |
|
AK314218.1 |
- |
2, 3 |
352 |
|
BC007225.1 |
- |
2, 3 |
335 |
|
BC011855.2 |
- |
2, 3 |
339 |
4139881 |
BC012343.1 |
- |
2, 3 |
331 |
4653507 |
BC014062.2 |
- |
2, 3 |
331 |
|
BC030145.2 |
- |
2, 3 |
331 |
|
BC032100.2 |
- |
2, 3 |
353 |
4660602 |
BX445320.2 |
- |
2, 3 |
415 |
|
DA172592.1 |
- |
2, 3 |
363 |
|
View Full Product Details