Gene Transcripts

Gene Symbol
Dgkd
Entrez Gene ID
368088
Gene Name
diacylglycerol kinase, delta
Gene Aliases
RGD1563309
Chromosome Location
Chr. 9: 95009638 - 95071531 on Build Rnor_6.0
UniGene ID
Rn.17832
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006245504.2 XP_006245566.2 10 1203
XM_006245505.2 XP_006245567.2 10 1203

View Full Product Details