Gene Transcripts

Gene Symbol
Mrgprb4
Entrez Gene ID
404658
Gene Name
MAS-related GPR, member B4
Gene Aliases
MrgB4
Chromosome Location
Chr. 1: 103425496 - 103426467 on Build Rnor_6.0
UniGene ID
Rn.155107
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002287.1 NP_001002287.1 1 574

View Full Product Details