Gene Transcripts

Gene Symbol
MTMR6
Entrez Gene ID
9107
Gene Name
myotubularin related protein 6
Gene Aliases
Chromosome Location
Chr. 13: 25246201 - 25287566 on Build GRCh38
UniGene ID
Hs.643702
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004685.3 NP_004676.3 12 1717
XM_011535306.1 XP_011533608.1 13 1777
XM_011535307.1 XP_011533609.1 13 1777
XM_017020845.1 XP_016876334.1 12 1663
XM_017020846.1 XP_016876335.1 9 982
GenBank mRNA AF072928.1 - 8 931
AF406619.1 - 12 1398
AK025507.1 - 2 107
AK314587.1 - 12 1582
AL832017.2 - 12 1640
BC040012.1 - 12 1589 5495564

View Full Product Details