Gene Transcripts

Gene Symbol
MTMR7
Entrez Gene ID
9108
Gene Name
myotubularin related protein 7
Gene Aliases
Chromosome Location
Chr. 8: 17296794 - 17413531 on Build GRCh38
UniGene ID
Hs.625674
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004686.4 NP_004677.3 14 1902
XM_006716414.3 XP_006716477.2 9 2268
XM_011544692.2 XP_011542994.1 11 1341
XM_011544693.2 XP_011542995.1 10 1803
XM_017013956.1 XP_016869445.1 13 1591
GenBank mRNA AK294468.1 - 14 1663
AK303762.1 - 6 725
AK303803.1 - 5 675
AK316022.1 - 14 1712
AK316283.1 - 14 1722
BC130577.1 - 14 1688 40146608
BC130579.1 - 14 1688 40146610
CR749240.1 - 14 1700

View Full Product Details