Gene Transcripts

Gene Symbol
Olr89
Entrez Gene ID
405012
Gene Name
olfactory receptor 89
Gene Aliases
Chromosome Location
Chr. 1: 168411463 - 168412413 on Build Rnor_6.0
UniGene ID
Rn.141897
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000745.1 NP_001000745.1 1 898

Gene Symbol
Olr88
Entrez Gene ID
405013
Gene Name
olfactory receptor 88
Gene Aliases
Chromosome Location
Chr. 1: 168403613 - 168404560 on Build Rnor_6.0
UniGene ID
Rn.141898
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000746.1 NP_001000746.1 1 895

View Full Product Details