Gene Transcripts
- Gene Symbol
- CCNB2
- Entrez Gene ID
- 9133
- Gene Name
- cyclin B2
- Gene Aliases
- HsT17299
- Chromosome Location
- Chr. 15: 59105085 - 59125045 on Build GRCh38
- UniGene ID
- Hs.194698
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004701.3 |
NP_004692.1
|
5, 6 |
774 |
|
GenBank mRNA |
AB020981.1 |
- |
5, 6 |
659 |
|
AF002822.1 |
- |
5, 6 |
609 |
|
AF087910.1 |
- |
5, 6 |
716 |
|
AK001404.1 |
- |
5, 6 |
738 |
|
AK222611.1 |
- |
5, 6 |
707 |
|
AL080146.1 |
- |
5, 6 |
727 |
|
AM851054.1 |
- |
5, 6 |
590 |
|
BC105086.1 |
- |
5, 6 |
722 |
8144089 |
BC105112.1 |
- |
5, 6 |
722 |
8144115 |
BT019458.1 |
- |
5, 6 |
590 |
|
CR533527.1 |
- |
5, 6 |
590 |
|
View Full Product Details