Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_017021760.1 | XP_016877249.1 | 27 | 2802 | |
XM_017021761.1 | XP_016877250.1 | 22 | 2255 | ||
GenBank mRNA | AF039687.1 | - | 6 | 829 | |
AK093783.1 | - | 12 | 1239 | ||
AK293236.1 | - | 27 | 2714 | ||
AK295010.1 | - | 27 | 2767 | ||
AK304695.1 | - | 27 | 2772 | ||
AK314281.1 | - | 10 | 978 | ||
BC020794.2 | - | 8 | 893 | ||
BC064364.1 | - | 8 | 885 | ||
BC107764.1 | - | 9 | 953 | ||
BX640804.1 | - | 4 | 458 | ||
BX640807.1 | - | 11 | 1095 | ||
BX648598.1 | - | 21 | 2172 |