Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004716.3 | NP_004707.2 | 6 | 1464 | |
GenBank mRNA | AK222907.1 | - | 6 | 1004 | |
AK312429.1 | - | 6 | 1039 | ||
BC006357.2 | - | 6 | 982 | 4125594 | |
BC010696.1 | - | 6 | 1034 | 3868257 | |
BT006870.1 | - | 4 | 833 | ||
U33849.1 | - | 6 | 984 | ||
U40623.1 | - | 4 | 854 |