Gene Transcripts

Gene Symbol
Smgc
Entrez Gene ID
446171
Gene Name
submandibular gland protein C
Gene Aliases
Chromosome Location
Chr. 7: 133048727 - 133078154 on Build Rnor_6.0
UniGene ID
Rn.81962
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004449.1 NP_001004449.1 2 143
GenBank mRNA AY459347.1 - 2 143

View Full Product Details