Gene Transcripts

Gene Symbol
Vom1r80
Entrez Gene ID
494238
Gene Name
vomeronasal 1 receptor 80
Gene Aliases
V1rc38
Chromosome Location
Chr. 4: 88026033 - 88026932 on Build Rnor_6.0
UniGene ID
Rn.129660
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009518.1 NP_001009518.1 1 443
GenBank mRNA AY510323.1 - 1 443

View Full Product Details