Gene Transcripts

Gene Symbol
Vom1r94
Entrez Gene ID
494265
Gene Name
vomeronasal 1 receptor 94
Gene Aliases
V1ra13, V1ra2
Chromosome Location
Chr. 4: 121810559 - 121811539 on Build Rnor_6.0
UniGene ID
Rn.129647
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008915.1 NP_001008915.1 1 855
GenBank mRNA AY510286.1 - 1 855

View Full Product Details