Gene Transcripts

Gene Symbol
Vom1r51
Entrez Gene ID
494281
Gene Name
vomeronasal 1 receptor 51
Gene Aliases
V1rm5
Chromosome Location
Chr. 1: 66872787 - 66873740 on Build Rnor_6.0
UniGene ID
Rn.129416
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008933.1 NP_001008933.1 1 201
GenBank mRNA AY510370.1 - 1 201

View Full Product Details