Gene Transcripts

Gene Symbol
Vom1r81
Entrez Gene ID
494313
Gene Name
vomeronasal 1 receptor 81
Gene Aliases
V1rc2
Chromosome Location
Chr. 4: 88048267 - 88049199 on Build Rnor_6.0
UniGene ID
Rn.129659
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008970.1 NP_001008970.1 1 454
GenBank mRNA AY510288.1 - 1 454

Gene Symbol
LOC100909473
Entrez Gene ID
100909473
Gene Name
vomeronasal type-1 receptor A16-like
Gene Aliases
Chromosome Location
Chr. Un|NW_007906889: 139649 - 140581 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006236570.1 XP_006236632.1 1 454

Gene Symbol
Vom1r75
Entrez Gene ID
494314
Gene Name
vomeronasal 1 receptor 75
Gene Aliases
V1rc29
Chromosome Location
Chr. 4: 87827582 - 87828514 on Build Rnor_6.0
UniGene ID
Rn.129665
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008972.1 NP_001008972.1 1 454
GenBank mRNA AY510283.1 - 1 454

View Full Product Details