Gene Transcripts

Gene Symbol
Uap1
Entrez Gene ID
498272
Gene Name
UDP-N-acteylglucosamine pyrophosphorylase 1
Gene Aliases
RGD1561967
Chromosome Location
Chr. 13: 88463532 - 88497901 on Build Rnor_6.0
UniGene ID
Rn.138943
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191930.1 NP_001178859.1 6, 7 1229
XM_006250222.1 XP_006250284.1 6, 7 1224
XM_006250223.1 XP_006250285.1 6, 7 1224

View Full Product Details