Gene Transcripts

Gene Symbol
Opn3
Entrez Gene ID
498289
Gene Name
opsin 3
Gene Aliases
Chromosome Location
Chr. 13: 93716481 - 93746994 on Build Rnor_6.0
UniGene ID
Rn.204549
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191933.1 NP_001178862.1 3 825

View Full Product Details