Gene Transcripts

Gene Symbol
Cpne2
Entrez Gene ID
498972
Gene Name
copine II
Gene Aliases
Chromosome Location
Chr. 19: 10860963 - 10898518 on Build Rnor_6.0
UniGene ID
Rn.161724
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256466.1 NP_001243395.1 3, 4 427
XM_008772319.1 XP_008770541.1 4, 5 604
GenBank mRNA AM747280.1 - 3, 4 427

View Full Product Details