Gene Transcripts

Gene Symbol
TRIP13
Entrez Gene ID
9319
Gene Name
thyroid hormone receptor interactor 13
Gene Aliases
16E1BP
Chromosome Location
Chr. 5: 892854 - 919348 on Build GRCh38
UniGene ID
Hs.436187
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166260.1 NP_001159732.1 3 434
NM_004237.3 NP_004228.1 3 434
XM_011514163.2 XP_011512465.1 3 404
GenBank mRNA AK310376.1 - 3 404
BC000404.2 - 3 399 2822981
BC019294.1 - 3 399 2822981
BG722761.1 - 3 436
CR456744.1 - 3 289
L40384.1 - 3 295
U96131.1 - 3 334

View Full Product Details