Gene Transcripts

Gene Symbol
Cks1b
Entrez Gene ID
499655
Gene Name
CDC28 protein kinase regulatory subunit 1B
Gene Aliases
RGD1561797
Chromosome Location
Chr. 2: 188740564 - 188745144 on Build Rnor_6.0
UniGene ID
Rn.119060
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135749.1 NP_001129221.1 3 951
GenBank mRNA BC167077.1 - 3 951
BC168780.1 - 3 982 6918892

View Full Product Details